Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Thermo Scientific™ Rheodyne™ Stators Available on GSA/VA Contract for Federal Government customers only.
Use these Thermo Scientific™ Rheodyne™ Stators for your Rheodyne Sample Injectors.
Manufacturer:  Thermo Scientific™ 9125043
 View more versions of this product
Catalog No. 03-052-387
Description
Description
Use Thermo Scientific™ Rheodyne™ Stators with popular Rheodyne injector models used in high-performance liquid chromatography (HPLC) systems.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Rheodyne™ injector models 9010/9030/9125
Rheodyne injector models 9010/9030/9125
Stators
For Rheodyne 9010/9030/9125
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.