Learn More
Raybiotech IncSupplier Diversity Partner Rhesus Monkey GASP-1 (WFIKKNRP) ELISA Kit DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner ELKGASP11
For Cell Culture Supernatants, Plasma, and Serum samples. Sensitivity: 49 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The antibody pair provided in this kit recognizes Rhesus Macaque GASP-1 (GPRASP1)
Catalog No. 50-194-9877