Learn More
Raybiotech IncSupplier Diversity Partner Rhesus Macaque uPAR ELISA Kit DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner ELKUPAR1
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 0.12 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Rhesus Macaque uPAR.
Catalog No. 50-194-9897