Learn More
Athens Research And Technology Inc RHEUMATOID FACTOR IGM HU 250UL
Supplier:  Athens Research And Technology Inc 1616180603250UL
Encompass_Preferred
Rheumatoid Factor (IgM), Human Plasma: 250 UL; Liquid in 20 mM Sodium Phosphate, pH 7.4, with 150 mM NaCl and 0.09% NaN3.; Elevation Level: = 1,000-2,000 IU/ml. Native; Purified from human plasma in the Athens Research & Technology laboratory. Immunology, Molecular Biology Reagent
Catalog No. 50-489-908