Learn More
RHO Mouse anti-Bovine, Human, Clone: B630, Abnova™
Mouse monoclonal antibody raised against native bovine RHO.
Manufacturer:  Abnova Corporation MAB12502
Catalog No. 89-934-857
Add to cart
Description
Description
Specifications
Specifications
RHO
B630
Mouse monoclonal antibody raised against native bovine RHO.
In PBS (10mM sodium azide)
RHO
Native purified RHO from bovine retinal outer segments.
Protein G purification
RUO
Primary
509933
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Immunocytochemistry Immunofluorescence (1:1000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Western Blot (1:5000) The optimal working dilution should be determined by the end user.
Liquid
Mouse
IgG1, kappa
50 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Bovine, Human