Learn More
Aviva Systems Biology Corp RHOA ANTIBODY 50UG/50UL
Supplier:  Aviva Systems Biology Corp ARP58521_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RHOA. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RHOA. Alternate names: ARH12, ARHA, RHO12, RHOH12ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-62340
Item Discontinued This item has been discontinued and is no longer available. Please reach out to your sales representative.