Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RhoA Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Supplier:  Thermo Scientific OSR00266W

Catalog No. PIOSR00266W

Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.


Description

Description

Glycerol (1:1) may be added for additional stability. Reconstitute in 100 μL of sterile water.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

RhoA
Polyclonal
RHOA
Arha, Arha2, ras homolog gene family member A, MGC72339
Rabbit
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Unconjugated
P61586, Q9QUI0, P61589
RHOA
A synthetic peptide from human Transforming protein RhoA conjugated to an immunogenic carrier protein
RUO
117273, 11848, 387
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit