Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RhoB Mouse anti-Human,Mouse, KHL, Clone: 9G5, MilliporeSigma™
Mouse Monoclonal Antibody
$177.00 - $444.00
Specifications
Antigen | RhoB |
---|---|
Clone | 9G5 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | KLH |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABS21782MI
|
MilliporeSigma
MABS217825UL |
25 μL |
Each for $177.00
|
|
|||||
MABS21781MI
|
MilliporeSigma
MABS2178100UL |
100 μL |
Each for $444.00
|
|
|||||
Description
RhoB Monoclonal antibody specifically detects RhoB in Human,Mouse samples. It is validated for Flow Cytometry.Specifications
RhoB | |
Flow Cytometry | |
KLH | |
Mouse | |
Human, Mouse | |
Rho-related GTP-binding protein RhoB;Rho cDNA clone 6;h6 | |
KLH-conjugated linear peptide corresponding to 19 amino acids from the C-terminal half of human Rho B. | |
Primary | |
Stable for 1 year at -20°C from date of receipt. | |
22.12 kDa calculated. |
9G5 | |
Monoclonal | |
Purified | |
Signaling | |
NP_004031.1 | |
RHOB, ARH6, ARHB | |
IgG2a κ | |
Protein G Purified | |
Clone 9G5 is a mouse monoclonal antibody that detects RhoB. it targets an epitope within 19 amino acids from the C-terminal half. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title