Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RHOBTB2 Mouse anti-Human, Clone: DBC2/3364, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP30748520UG
Description
RHOBTB2 Monoclonal specifically detects RHOBTB2 in Human samples. It is validated for Immunohistochemistry-Paraffin, Protein Array.Specifications
RHOBTB2 | |
Monoclonal | |
0.2 mg/ml | |
Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
DBC2KIAA0717Deleted in breast cancer 2 gene protein, p83, Rho-related BTB domain containing 2, rho-related BTB domain-containing protein 2 | |
Human recombinant RHOBTB2 protein fragment (around aa 554-604) (exact sequence is proprietary) | |
20 μg | |
Signal Transduction | |
23221 | |
Store at 4°C. | |
IgG2c κ |
Immunohistochemistry (Paraffin), Peptide Array | |
DBC2/3364 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction