Learn More
RhoBTB3 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA551047
Catalog No. PA551047
Add to cart
Description
Description
RHOBTB3 Polyclonal Antibody detects endogenous levels of RHOBTB3 protein.
Rab9-regulated ATPase required for endosome to Golgi transport. Involved in transport vesicle docking at the Golgi complex, possibly by participating in release M6PRBP1/TIP47 from vesicles to permit their efficient docking and fusion at the Golgi. Specifically binds Rab9, but not other Rab proteins. Has low intrinsic ATPase activity due to autoinhibition, which is relieved by Rab9. {ECO:0000269; PubMed:19490898}.
Specifications
Specifications
RHOBTB3
Polyclonal
Unconjugated
RHOBTB3
Liquid
RHOBTB3
Synthesized peptide derived from the Internal region of human RHOBTB3.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
ELISA, Immunocytochemistry, Immunohistochemistry, Western Blot
1 mg/mL
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
O94955, Q9CTN4
KIAA0878
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
22836, 73296