Learn More
RHOC Mouse anti-Human, Clone: OTI6B4, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF806449
Catalog No. 50-166-7841
Add to cart
Description
Description
Synthetic peptide sequence is VFEMATRAGLQVRKNKRRRG Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Blood-group antigens are generally defined as molecules formed by sequential addition of saccharides to the carbohydrate side chains of lipids and proteins detected on erythrocytes and certain epithelial cells. The A, B and H antigens are reported to undergo modulation during maligt cellular transformation. Blood group related antigens represent a group of carbohydrate determits carried on both glycolipids and glycoproteins. They are usually mucin-type, and are detected on erythrocytes, certain epithelial cells, and in secretions of certain individuals. Sixteen genetically and biosynthetically distinct but inter-related specificities belong to this group of antigens, including A, B, H, Lewis A, Lewis B, Lewis X, Lewis Y, and precursor type 1 chain antigens.
Specifications
Specifications
RHOC
Monoclonal
Unconjugated
RHOC
Lyophilized
RHOC
Synthetic peptide corresponding to C terminal of human RHOC.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI6B4
PBS with 8% trehalose and no preservative; pH 7.3
P08134
ARH9, ARHC, H9, RHOH9
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
389