Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Rhodamine 110 bis(Z-L-isoleucyl-L-prolyl-L-arginine amide) dihydrochloride

Click to view available options
Quantity:
0.5mg
1mg
MFCD00467932
(Z-Ile-Pro-Arg)2-R110; BZiPAR
MFCD00467932
(Z-Ile-Pro-Arg)2-R110; BZiPAR
Quantity 0.5mg
Color White
Formula Weight 1404.46
Physical Form Solid
Chemical Name or Material Rhodamine 110 bis(Z-L-isoleucyl-L-prolyl-L-arginine amide) dihydrochloride

storageNote1 : -30° to -10°C