Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Rhodamine 6G perchlorate, 96%

Catalog No. p-7032237
Click to view available options
Quantity:
250 mg
1 g

CAS: 13161-28-9 | C28H31ClN2O7 | 543.013 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13161-28-9
Molecular Formula C28H31ClN2O7
Molecular Weight (g/mol) 543.013
MDL Number MFCD00013117
InChI Key HDAFVOZRAUFNQH-UHFFFAOYSA-N
Synonym rhodamine 6g perchlorate, rhodamine 6g perchlorate, dye content 99 %, ethyl 2-6e-3-ethylamino-6-ethylimino-2,7-dimethylxanthen-9-yl benzoate; perchloric acid, ethyl 2-6z-3-ethylamino-6-ethylimino-2,7-dimethylxanthen-9-yl benzoate; perchloric acid
PubChem CID 13651880
IUPAC Name ethyl 2-[3-(ethylamino)-6-ethylimino-2,7-dimethylxanthen-9-yl]benzoate;perchloric acid
SMILES CCNC1=C(C=C2C(=C1)OC3=CC(=NCC)C(=CC3=C2C4=CC=CC=C4C(=O)OCC)C)C.OCl(=O)(=O)=O

Specifications

Melting Point >240°C
Quantity 250 mg
Formula Weight 543.02
Percent Purity 96%
Chemical Name or Material Rhodamine 6G perchlorate
Hazard Category H302-H315-H319-H335-H500
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 236-099-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.