Learn More
Aat BioquestSupplier Diversity Partner Rhodamine 800 *CAS 137993-41-0* DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 68
Rh 800 was originally developed as a laser dye. It is one of the few rhodamines that have near infrared fluorescence. It is also used for staining mitochondria.
Catalog No. 50-195-2692