Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Rhodanine-3-acetic acid, 98%

Click to view available options
Quantity:
5g
25g
100g
5718-83-2
C5H5NO3S2
191.219
MFCD00005491
JGRMXPSUZIYDRR-UHFFFAOYSA-N
rhodanine-3-acetic acid,rhodanine-n-acetic acid,n-carboxymethylrhodanine,3-rhodanineacetic acid,3-thiazolidineacetic acid, 4-oxo-2-thioxo,3-carboxymethyl rhodanine,n-carboxymethyl rhodanine,2-4-oxo-2-thioxothiazolidin-3-yl acetic acid,4-oxo-2-thioxo-3-thiazolidineacetic acid,4-oxo-2-thioxo-3-thiazolidinylacetic acid
79793
2-(4-oxo-2-sulfanylidene-1,3-thiazolidin-3-yl)acetic acid
C1C(=O)N(C(=S)S1)CC(=O)O
5718-83-2
C5H5NO3S2
191.219
MFCD00005491
JGRMXPSUZIYDRR-UHFFFAOYSA-N
rhodanine-3-acetic acid,rhodanine-n-acetic acid,n-carboxymethylrhodanine,3-rhodanineacetic acid,3-thiazolidineacetic acid, 4-oxo-2-thioxo,3-carboxymethyl rhodanine,n-carboxymethyl rhodanine,2-4-oxo-2-thioxothiazolidin-3-yl acetic acid,4-oxo-2-thioxo-3-thiazolidineacetic acid,4-oxo-2-thioxo-3-thiazolidinylacetic acid
79793
2-(4-oxo-2-sulfanylidene-1,3-thiazolidin-3-yl)acetic acid
C1C(=O)N(C(=S)S1)CC(=O)O
Quantity 100g
Melting Point 145°C to 148°C
Beilstein 149513
Formula Weight 191.23
Percent Purity 98%
Chemical Name or Material Rhodanine-3-acetic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA