Alfa Aesar™ Rhodium, 1% on 2.5mm alumina spheres

Manufacturer: Alfa Aesar™ 47040-30

Quantity 250g

 View more versions of this product

Catalog No. AA4704030

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 7440-16-6
    MDL Number MFCD00011201
    Chemical Name or Material Rhenium, 1% on alumina spheres, 2.5mm alumina spheres

    Specifications

    Physical Form Spheres
    CAS 7440-16-6
    TSCA TSCA
    MDL Number MFCD00011201
    Quantity 250g
    Chemical Name or Material Rhenium, 1% on alumina spheres, 2.5mm alumina spheres

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit