Alfa Aesar™ Rhodium, 1% on alumina powder, reduced

Manufacturer: Alfa Aesar™ 1176909

Quantity 10g

 View more versions of this product

Catalog No. AA1176909

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-16-6
    MDL Number MFCD00011201
    Chemical Name or Material Rhodium, 1% on alumina powder, Reduced

    Specifications

    Health Hazard 3 GHS P Statement
    P261-P304+P340-P312-P405-P403+P233-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Store locked up.
    Store in a well-ventilated place.
    Keep container tightly closed.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H335
    May cause respiratory irritation.
    TSCA TSCA
    MDL Number MFCD00011201
    CAS 7440-16-6
    Health Hazard 1 Warning
    Quantity 10g
    Chemical Name or Material Rhodium, 1% on alumina powder, Reduced

    GHS H Statement
    H335
    May cause respiratory irritation.

    GHS P Statement
    P261-P304+P340-P312-P405-P403+P233-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Store locked up.
    Store in a well-ventilated place.
    Keep container tightly closed.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit