Alfa Aesar™ Rhodium, 5% on carbon paste, Type 594

Manufacturer: Alfa Aesar™ H3741014

Quantity 25g

 View more versions of this product

Catalog No. AAH3741014

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-16-6
    MDL Number MFCD00011201
    Chemical Name or Material Rhodium, 5% on carbon paste, Type 594

    Specifications

    Physical Form Solid
    MDL Number MFCD00011201
    CAS 7440-16-6
    Color Dark gray to black
    Quantity 25g
    TSCA TSCA
    Odor Odorless
    Chemical Name or Material Rhodium, 5% on carbon paste, Type 594

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit