Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rhodium on alumina, 5% Rh, powder, ACROS Organics™

Click to view available options
Quantity:
5g
25g
7440-16-6,1344-28-1
Rh
102.906
MFCD00011201
MHOVAHRLVXNVSD-UHFFFAOYSA-N
fume,on carbon,rhodium, elemental,rodio,black,rhodium, metal,unii-dmk383dsac,on alumina,rh/al2o3,dmk383dsac
23948
CHEBI:33359
rhodium
[Rh]
7440-16-6,1344-28-1
Rh
102.906
MFCD00011201
MHOVAHRLVXNVSD-UHFFFAOYSA-N
fume,on carbon,rhodium, elemental,rodio,black,rhodium, metal,unii-dmk383dsac,on alumina,rh/al2o3,dmk383dsac
23948
CHEBI:33359
rhodium
[Rh]
Quantity 25g
Packaging Glass bottle
Formula Weight 102.91
Melting Point 2072.0°C
Boiling Point 3147.0°C
Chemical Name or Material Rhodium on alumina