Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rhodium(II) trifluoroacetate dimer, Acros Organics™

Click to view available options
Quantity:
250mg
1g
Packaging:
Glass bottle
31126-95-1
C8F12O8Rh2
657.88
MFCD00209611
SZQVMUPTZFMHQT-UHFFFAOYSA-N
rhodium ii trifluoroacetate dimer,trifluoroacetic acid rhodium ii salt dimer,dirhodium ii tetra trifluoroacetate,rhodium ii trifluoroacetate dimer, ?rh2 cf3coo 4,bis 2,2,2-trifluoroacetyl oxy rhodio 2,2,2-trifluoroacetate
10985187
rhodium;2,2,2-trifluoroacetic acid
C(=O)(C(F)(F)F)O.C(=O)(C(F)(F)F)O.C(=O)(C(F)(F)F)O.C(=O)(C(F)(F)F)O.[Rh].[Rh]
31126-95-1
C8F12O8Rh2
657.88
MFCD00209611
SZQVMUPTZFMHQT-UHFFFAOYSA-N
rhodium ii trifluoroacetate dimer,trifluoroacetic acid rhodium ii salt dimer,dirhodium ii tetra trifluoroacetate,rhodium ii trifluoroacetate dimer, ?rh2 cf3coo 4,bis 2,2,2-trifluoroacetyl oxy rhodio 2,2,2-trifluoroacetate
10985187
rhodium;2,2,2-trifluoroacetic acid
C(=O)(C(F)(F)F)O.C(=O)(C(F)(F)F)O.C(=O)(C(F)(F)F)O.C(=O)(C(F)(F)F)O.[Rh].[Rh]
Formula Weight 657.88
Physical Form Crystalline Powder
Infrared Spectrum Authentic
Packaging Glass bottle
Rhodium (Rh) 31.0% min.
Color Green
Quantity 250mg
Chemical Name or Material Rhodium(II) trifluoroacetate dimer

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning