Learn More
Rhodium(II) acetate, dimer, Premion™, 99.99% (metals basis), Rh 46.2% min, Thermo Scientific Chemicals
Description
Catalyst for cyclopropanation of alkenes, oxidation of alcohols, cyclization reactions involving -diazo carbonyl groups, insertion into C-H and X-H bonds (X-H is NH or SH or OH) and for ylide formation. More reactive and useful in differentiating ribonucleosides and deoxynucleosides. It is also used in functionalizing fullerenes into polymers. Efficient catalyst for hydrogen transfer from 2-propanol to cyclohexanone and other unsaturated compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 205°C |
Color | Green |
Physical Form | Powder |
Quantity | 0.25 g |
Assay Percent Range | 99.99% (metals basis) |
Solubility Information | Soluble in aqueous solution like water. Slightly soluble in ethanol |
IUPAC Name | rhodium(2+);tetraacetate |
Formula Weight | 441.99 |
Odor | Odorless |
Appearance | Green powder |
Show More |
Safety and Handling
Hazard Category | H315-H319 |
Precautionary Statement | P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362 |
EINECSNumber | 240-084-8 |
RTECSNumber | VI9361000 |
TSCA | No |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.