Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rhodium(II) acetate, dimer, 99.99% (metals basis), Rh 46.2% min., Alfa Aesar™ Premion™

For Use in Cyclopropanation of Alkenes

Manufacturer:  Alfa Aesar 1056006

Quantity 5g

 View more versions of this product

Catalog No. AA1056006


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

15956-28-2
99.99% (metals basis)
441.99
Tetrakis(aceto)dirhodium(II)
Powder
C8H12O8Rh2
MFCD00003538
Rhodium(II) acetate, dimer, Rh 46.2% min.

Specifications

15956-28-2
99.99% (metals basis)
Green powder
MFCD00003538
Odorless
5g
Powder
C8H12O8Rh2
441.99
Tetrakis(aceto)dirhodium(II)
Green
Rhodium(II) acetate, dimer, Rh 46.2% min.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit