Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rhodium(II) acetate dimer, dihydrate, ca 43% Rh, ACROS Organics™

Click to view available options
Quantity:
250mg
1g
Packaging:
Glass bottle
29998-99-0
C8H16O10Rh2
478.02
MFCD00149212
Tetrakis[micron-(acetato-O:O')]diaquadirhodium
29998-99-0
C8H16O10Rh2
478.02
MFCD00149212
Tetrakis[micron-(acetato-O:O')]diaquadirhodium
Linear Formula [(CH3CO2)2RhH2O]2
Fieser 08,434; 13,266; 15,278; 16,289; 17,298; 05,571
Solubility Information Solubility in water: insoluble. Other solubilities: moderately soluble in many polar solvents like eth,anol,acetone,acetic acid,acetonitrile,dmso,ni,tromethane,thf,triethyl phosphate
Formula Weight 478.02
Physical Form Fine Powder
Percent Purity ca 43% Rh
Infrared Spectrum Authentic
Packaging Glass bottle
Solubility Solubility in water: insoluble. Other solubilities: moderately soluble in many polar solvents like eth, anol, acetone, acetic acid, acetonitrile, dmso, ni, tromethane, thf, triethyl phosphate.
Color Green
Quantity 1g
Chemical Name or Material Rhodium(II) acetate dimer, dihydrate, ca 43% Rh
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning