Alfa Aesar™ Rhodium(III) 2,4-pentanedionate, Premion™, 99.99% (metals basis), Rh 25.2% min

Manufacturer: Alfa Aesar™ 1056103

Quantity 1g

 View more versions of this product

Catalog No. AA1056103

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 14284-92-5
    Formula Weight 400.23
    MDL Number MFCD00083144
    Synonym Rhodium(III) acetylacetonate
    Chemical Name or Material Rhodium(III) 2,4-pentanedionate
    Grade Premion™

    Specifications

    Formula Weight 400.23
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    MDL Number MFCD00083144
    Health Hazard 1 Warning
    TSCA TSCA
    Physical Form Crystalline
    CAS 14284-92-5
    Synonym Rhodium(III) acetylacetonate
    Quantity 1g
    Solubility Information Very soluble in chloroform; Soluble in alcohol
    Concentration Rh 25.2% min.
    Melting Point 263° to 264°C
    Chemical Name or Material Rhodium(III) 2,4-pentanedionate
    Assay 99.99%, (metals basis)
    Grade Premion™

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit