Alfa Aesar™ Rhodium(III) chloride, anhydrous, 99.9% (metals basis), Rh 48.7% min

Manufacturer: Alfa Aesar™ 1181503

Quantity 1g

 View more versions of this product

Catalog No. AA1181503

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10049-07-7
    Linear Formula RhCl3
    Formula Weight 209.26
    MDL Number MFCD00011204
    Merck Index 14,8188
    Chemical Name or Material Rhodium(III) chloride, Anhydrous

    Specifications

    Formula Weight 209.26
    Health Hazard 3 GHS P Statement
    P280-P281-P305+P351+P338-P310-P405-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Powder
    Linear Formula RhCl3
    Health Hazard 1 Danger
    Odor Odorless
    Sensitivity Hygroscopic
    MDL Number MFCD00011204
    CAS 10049-07-7
    Merck Index 14,8188
    Color Red-brown
    Concentration Rh 48.7% min.
    TSCA TSCA
    Health Hazard 2 GHS H Statement
    H318-H290-H341-H302
    Causes serious eye damage.
    May be corrosive to metals.
    Suspected of causing genetic defects.
    Harmful if swallowed.
    RTECS Number VI9275000
    Quantity 1g
    Chemical Name or Material Rhodium(III) chloride, Anhydrous
    Assay 99.9%, (metals basis)

    GHS H Statement
    H318-H290-H341-H302
    Causes serious eye damage.
    May be corrosive to metals.
    Suspected of causing genetic defects.
    Harmful if swallowed.

    GHS P Statement
    P280-P281-P305+P351+P338-P310-P405-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit