Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Rhodium(III) chloride, anhydrous, 99.9% (metals basis), Rh 48.7% min

Manufacturer:  Alfa Aesar™ 1181506

Quantity 5g

 View more versions of this product

Catalog No. AA1181506


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

10049-07-7
209.26
14,8188
RhCl3
MFCD00011204
Rhodium(III) chloride, Anhydrous

Specifications

10049-07-7
RhCl3
MFCD00011204
Rh 48.7% min.
Hygroscopic
5g
99.9%, (metals basis)
Powder
209.26
14,8188
Odorless
Red-brown
Rhodium(III) chloride, Anhydrous

Safety and Handling

SDS & Safety Documents

GHS H Statement
H318-H290-H341-H302
Causes serious eye damage.
May be corrosive to metals.
Suspected of causing genetic defects.
Harmful if swallowed.

GHS P Statement
P280-P281-P305+P351+P338-P310-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : VI9275000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit