Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Rhodium(III) chloride, anhydrous, 99.9% (metals basis), Rh 48.7% min

Click to view available options
Quantity:
0.25g
1g
5g
10049-07-7
MFCD00011204
10049-07-7
MFCD00011204
Linear Formula RhCl3
Merck Index 14,8188
Solubility Information Insoluble in water. Soluble in hydroxide and cyanide solutions.
Formula Weight 209.26
Physical Form Powder
Percent Purity 99.9%
Concentration Rh 48.7% min.
Odor Odorless
Sensitivity Hygroscopic
Color Red to Brown
Quantity 0.25g
Chemical Name or Material Rhodium(III) chloride, Anhydrous
Assay (metals basis)
Show More Show Less

GHS H Statement
H318-H290-H341-H302
Causes serious eye damage.
May be corrosive to metals.
Suspected of causing genetic defects.
Harmful if swallowed.

P201-P202-P234-P264b-P270-P281-P301+P312-P305+P351+P338-P308+P313-P310-P330-P390-P501c

H290-H302-H318-H341

DOTInformation : Hazard Class: 8; Packaging Group: III

EINECSNumber : 233-165-4

RTECSNumber : VI9275000

TSCA : Yes

Recommended Storage : Ambient temperatures