Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rhodium(III) chloride hydrate, 38% Rh, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
20765-98-4
Cl3Rh·xH2O
209.26
20765-98-4
Cl3Rh·xH2O
209.26
Linear Formula RhCl3·xH2O
Fieser 10,343; 11,460; 14,271; 16,292; 17,366
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alchol and hcl,insoluble in ether
Formula Weight 209.26
Physical Form Crystalline Powder
Percent Purity 38% Rh
Packaging Glass bottle
Solubility Solubility in water: soluble. Other solubilities: soluble in alchol and hcl, insoluble in ether
Color Brown to Brown-Red
Melting Point 100.0°C
Quantity 5g
Chemical Name or Material Rhodium(III) chloride hydrate
Show More Show Less

GHS H Statement
Causes serious eye damage.
Harmful if swallowed.
Suspected of causing genetic defects.
May be corrosive to metals.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately

GHS Signal Word: Danger