Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rhodium(III) chloride hydrate, Rh 38.5-45.5%, Alfa Aesar™

Manufacturer:  Alfa Aesar 1103206

Quantity 5g

 View more versions of this product

Catalog No. AA1103206


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

20765-98-4
38.5 to 45.5%
209.26 (anhydrous)
Rhodium trichloride hydrate
Crystalline
RhCl3·xH2O
MFCD00149839
Rhodium(III) chloride hydrate, Rh 38.5-45.5%

Specifications

20765-98-4
38.5 to 45.5%
Dark red crystals
MFCD00149839
Dark red
Rhodium(III) chloride hydrate, Rh 38.5-45.5%
Crystalline
RhCl3·xH2O
209.26 (anhydrous)
Rhodium trichloride hydrate
5g

Safety and Handling

SDS & Safety Documents

In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. This material and/or its container must be disposed of as hazardous waste.

H318-H290-H341-H302: Causes serious eye damage. May be corrosive to metals. Suspected of causing genetic defects. Harmful if swallowed.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit