Alfa Aesar™ Rhodium(III) oxide, anhydrous, 99.9% (metals basis), Rh 80.6% min

Manufacturer: Alfa Aesar™ 1181404

Quantity 2g

 View more versions of this product

Catalog No. AA1181404

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12036-35-0
    Formula Weight 253.81
    MDL Number MFCD00011205
    Chemical Name or Material Rhodium(III) oxide, Anhydrous

    Specifications

    Formula Weight 253.81
    Odor Odorless
    Physical Form Powder
    Sensitivity Hygroscopic
    Color Gray to black
    Melting Point 1100°C (dec.)
    Quantity 2g
    Concentration Rh 80.6% min.
    Density 8.20g/mL
    TSCA TSCA
    MDL Number MFCD00011205
    CAS 12036-35-0
    Chemical Name or Material Rhodium(III) oxide, Anhydrous
    Assay 99.9%, (metals basis)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit