Alfa Aesar™ Rhodium(III) sulfate, bright plating solution, Rh 5% (cont. Rh)

Manufacturer: Alfa Aesar™ 12635NA

Quantity 2g

 View more versions of this product

Catalog No. AA12635NA

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10489-46-0
    Linear Formula Rh2(SO4)3
    Formula Weight 493.99
    MDL Number MFCD00016309
    UN Number UN3264
    Chemical Name or Material Rhodium(III) sulfate, Bright plating solution

    Specifications

    Formula Weight 493.99
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H314
    Causes severe skin burns and eye damage.
    Color Red-orange
    Physical Form Liquid
    UN Number UN3264
    Health Hazard 1 Danger
    RTECS Number FF9335000
    TSCA TSCA
    Density 1.4g/mL
    MDL Number MFCD00016309
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.
    Quantity 2g
    Concentration Rh 5% (cont. Rh)
    Linear Formula Rh2(SO4)3
    CAS 10489-46-0
    Odor Acidic
    Chemical Name or Material Rhodium(III) sulfate, Bright plating solution

    GHS H Statement
    H314
    Causes severe skin burns and eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit