Alfa Aesar™ Rhodium(III) sulfate tetrahydrate, Premion™, 99.99% (metals basis), Rh 35.9% min

Manufacturer: Alfa Aesar™ 4101606

Quantity 5g

 View more versions of this product

Catalog No. AA4101606

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 15274-78-9
    Linear Formula Rh2(SO4)3·4H2O
    Formula Weight 566.05 (493.99 Anhydrous)
    MDL Number MFCD00798544
    UN Number UN3261
    Chemical Name or Material Rhodium(III) sulfate tetrahydrate
    Grade Premion™

    Specifications

    Formula Weight 566.05 (493.99 Anhydrous)
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H314-H318-H290
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    May be corrosive to metals.
    Linear Formula Rh2(SO4)3·4H2O
    MDL Number MFCD00798544
    UN Number UN3261
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
    Odor Odorless
    Physical Form Crystalline
    Color Brown
    TSCA TSCA
    CAS 15274-78-9
    Quantity 5g
    Concentration Rh 35.9% min.
    Health Hazard 1 Danger
    Chemical Name or Material Rhodium(III) sulfate tetrahydrate
    Assay 99.99%, (metals basis)
    Grade Premion™

    GHS H Statement
    H314-H318-H290
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    May be corrosive to metals.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit