Rhodium, plasma standard solution, Rh 1000µg/mL, Alfa Aesar™, Specpure™

Manufacturer: Alfa Aesar 35754AE

Quantity 100mL

 View more versions of this product

Catalog No. AA35754AE

  Check Availability
Add to Cart

    Chemical Identifiers

    Physical Form Liquid
    Molecular Formula RhCl3 in 20% HCl
    MDL Number MFCD00011201
    Chemical Name or Material Rhodium, Rh 1000μg/mL

    Specifications

    Quantity 100mL
    DOT Information DOT Class: 8; Packing Group: II
    Health Hazard 2 Keep container in a well ventilated place. Keep away from combustible material. When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.
    EINECS Number 231-595-7
    Physical Form Liquid
    Appearance Liquid
    Molecular Formula RhCl3 in 20% HCl
    MDL Number MFCD00011201
    Health Hazard 1 H314-H335: Causes severe skin burns and eye damage. May cause respiratory irritation.
    Odor Characteristic
    Chemical Name or Material Rhodium, Rh 1000μg/mL

    Keep container in a well ventilated place. Keep away from combustible material. When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.

    H314-H335: Causes severe skin burns and eye damage. May cause respiratory irritation.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit