Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Rhodium, plasma standard solution, Specpure™, Rh 10,000μg/mL
Click to view available options
Quantity:
50mL
100mL
500mL
Specifications
UN Number | UN1789 |
Concentration | RhCl3 in 20% HCl |
Solubility Information | Miscible with water. |
Physical Form | Liquid |
Grade | Specpure™ |
Quantity | 500mL |
Chemical Name or Material | Rhodium plasma standard solution, Rh 10000μg/mL |
Safety and Handling
GHS H Statement
H314-H335
Causes severe skin burns and eye damage.
May cause respiratory irritation.
P201-P202-P234-P260-P264b-P271-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
H290-H314-H331-H335-H341
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: HYDROCHLORIC ACID
EINECSNumber : 231-595-7
TSCA : Yes
Recommended Storage : Ambient temperatures