Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Rhodium powder, -22 mesh, Premion™, 99.97% (metals basis), precious metals typically <20ppm each

Click to view available options
Quantity:
0.25g
1g
7440-16-6
Rh
102.906
MFCD00011201
MHOVAHRLVXNVSD-UHFFFAOYSA-N
fume,on carbon,rhodium, elemental,rodio,black,rhodium, metal,unii-dmk383dsac,on alumina,rh/al2o3,dmk383dsac
23948
CHEBI:33359
rhodium
[Rh]
7440-16-6
Rh
102.906
MFCD00011201
MHOVAHRLVXNVSD-UHFFFAOYSA-N
fume,on carbon,rhodium, elemental,rodio,black,rhodium, metal,unii-dmk383dsac,on alumina,rh/al2o3,dmk383dsac
23948
CHEBI:33359
rhodium
[Rh]
UN Number UN3089
Physical Form Powder
Percent Purity 99.97%
Grade Premion™
Mesh Size −22 Mesh
Odor Odorless
Color Gray
Quantity 0.25g
Chemical Name or Material Rhodium powder, Precious metals typically <20ppm each
Assay (metals basis)

GHS H Statement
H228
Flammable solid.

GHS P Statement
P210-P280-P240-P241-P370+P378a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/ equipment.
In case of fire: Use for extinction: CO2, powder or water spray.

H500

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.

EINECSNumber : 231-125-0

RTECSNumber : VI9069000

TSCA : Yes

Recommended Storage : Ambient temperatures