Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rhodium standard solution, for AAS, 1mg/mL Rh in 10% HCl, ACROS Organics™

Click to view available options
Quantity:
100mL
Packaging:
Plastic bottle
7440-16-6,7647-01-0,7732-18-5,13569-65-8
Rh
102.906
MFCD00011201
MHOVAHRLVXNVSD-UHFFFAOYSA-N
23948
CHEBI:33359
rhodium
[Rh]
7440-16-6,7647-01-0,7732-18-5,13569-65-8
Rh
102.906
MFCD00011201
MHOVAHRLVXNVSD-UHFFFAOYSA-N
23948
CHEBI:33359
rhodium
[Rh]
for atomic adsorption spectroscopy
Concentration 997 to 1003 μg/mL of Rh (3+) in 10% HCl (20°C)
Formula Weight 102.91
Physical Form Liquid
Grade AAS
Packaging Plastic bottle
Color Red
Quantity 100mL
Chemical Name or Material Rhodium standard solution, 1 mg/ml Rh in 10% HCl

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May be corrosive to metals.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Keep only in original container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning