Learn More
High Purity StandardsSupplier Diversity Partner RHODIUM STNDRD 100ML 10%HCI DFS Item
Manufacturer:  High Purity StandardsSupplier Diversity Partner 1000442100
1000 ug/mL Rhodium in 10% HCl, 100mL, sourced from RhCl3, 18 mo expiry date. This standard is traceable to NIST and certified Guide 34 and 17025.
Catalog No. NC0088387