Alfa Aesar™ Rhodium wire, 0.5mm (0.02 in.) dia., 99.8% (metals basis)

Manufacturer: Alfa Aesar™ 11576BS

Quantity 10cm

 View more versions of this product

Catalog No. AA11576BS

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-16-6
    MDL Number MFCD00011201
    Chemical Name or Material Rhodium wire, 0.5mm (0.02 in.) dia.

    Specifications

    Physical Form Wire
    MDL Number MFCD00011201
    CAS 7440-16-6
    Weight ≈2.44g/m
    Quantity 10cm
    TSCA TSCA
    Chemical Name or Material Rhodium wire, 0.5mm (0.02 in.) dia.
    Assay 99.8%, (metals basis)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit