Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rhodopsin Mouse anti-Amphibian, Avian, Fish, Mammalia, Clone: 4D2, Novus Biologicals™

Mouse MonoclonalAntibody

$199.00 - $355.50

Specifications

Antigen Rhodopsin
Clone 4D2
Dilution Western Blot 1:1000, ELISA, Immunohistochemistry 1000, Immunocytochemistry/Immunofluorescence, Immunoprecipitation
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Conjugate Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Conjugate Price Quantity & Availability  
NB59690100U View Documents Novus BiologicalsSupplier Diversity Partner
NBP259690100UG
100 ug Unconjugated Each for $355.50
Add to cart
 
NB5969025UG View Documents Novus BiologicalsSupplier Diversity Partner
NBP25969025UG
25 ug Unconjugated Each for $199.00
Add to cart
 
Description

Description

Rhodopsin Monoclonal antibody specifically detects Rhodopsin in Amphibian, Avian, Fish, Mammal samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications

Specifications

Rhodopsin
Western Blot 1:1000, ELISA, Immunohistochemistry 1000, Immunocytochemistry/Immunofluorescence, Immunoprecipitation
Purified
Amphibian, Avian, Fish, Mammalia
PBS pH7.4, 50% glycerol with 0.09% Sodium Azide
Bovine Rhodopsin
Monoclonal
Protein G purified
Binds specifically to the N-terminus of Rhodopsin. Does not detect Rhodopsin in invertebrates.
4D2
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Mouse
RUO
6010
IgG1
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.

For Research Use Only (RUO)

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit