Learn More
Rhodopsin Mouse anti-Human, Mouse, Bovine, Vertebrate, DyLight 550, Clone: 1D4, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP147602R
Catalog No. NBP147602R
Add to cart
Description
Description
Rhodopsin Monoclonal antibody specifically detects Rhodopsin in Human, Mouse, Bovine, Vertebrate samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).
Specifications
Specifications
Rhodopsin
1D4
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Purified
RHO
Bovine Rhodopsin
Protein G purified
GPCR, Phospho Specific, Signal Transduction, Vision
Primary
6010
Bovine, Human, Mouse, Vertebrate
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
DyLight 550
50mM Sodium Borate with 0.05% Sodium Azide
CSNBAD1, MGC138309, OPN2MGC138311, opsin 2, rod pigment, opsin-2, retinitis pigmentosa 4, autosomal dominant, rhodopsin, RP4
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Detects ~40kDa. Recognizes Rhodopsin (native and recombinant forms). No known reactivity to other proteins. Binds specifically to the C-terminal epitope -T-E-T-S-Q-V-A-P-A-(COOH).