Learn More
Epredia™ Lab Vision™ Rhodopsin (Opsin) Ab-1 Mouse Monoclonal Antibody, 200μg/mL, biotinylated, BSA and azide
Provide accurate, reproducible results with the Epredia™ Rhodopsin (Opsin) Ab-1, Mouse Monoclonal Antibody.
$679.00
Specifications
Antigen | Rhodopsin (Opsin) Ab-1 |
---|---|
Clone | RET-P1 |
Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Host Species | Mouse |
Catalog Number | Mfr. No. | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Form | Quantity | Price | Quantity & Availability | ||||
MS1233P1
|
Epredia™
MS1233P1 |
Concentrated | 500 μL |
Each for $679.00
|
|
||||
MS1233B0
|
Epredia™
MS1233B0 |
Concentrated | 100 μL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
MS1233P0
|
Epredia™
MS1233P0 |
Concentrated | 100 μL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
MS1233P1ABX
|
Epredia™
MS1233P1ABX |
Concentrated | 100 μL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
MS1233PABX
|
Epredia™
MS1233PABX |
Concentrated | 200 μL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
MS1233B1
|
Epredia™
MS1233B1 |
Concentrated | 500 μL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
MS1233B
|
Epredia™
MS1233B |
Concentrated | 1 mL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
MS1233P
|
Epredia™
MS1233P |
Concentrated | 1 mL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
MS1233R7
|
Epredia™
MS1233R7 |
Prediluted | 7 mL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
Description
Rhodopsin is a glycoprotein which is a visual pigment in mammals, birds, amphibians, and reptiles. The rhodopsin protein is present on the photoreceptor cells in the retina.Host Species: Mouse
Clone: RET-P1
Isotype: IgG1
Species Reactivity: Human, Mouse and Rat. Others not known.
Epitope: Not determined
Immunogen: Membrane preparation from adult rat retina
Molecular Weight: 39kDa
Positive Control: Brain or retina
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
Note:
USA: RUO; Int'l: RUO
Specifications
Rhodopsin (Opsin) Ab-1 | |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Mouse | |
Cancer and Tumor Biology | |
Membrane preparation from adult rat retina | |
Primary |
RET-P1 | |
Monoclonal | |
RUO | |
Human, Mouse, Rat | |
IgG1 |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.