Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rhodopsin, Rabbit anti-Human,Mouse,Rat,Amphibian,Bovine,Zebrafish, Clone: Rho 1D4, Novus Biologicals™

Rabbit Monoclonal Antibody

$156.50 - $419.00

Specifications

Antigen Rhodopsin
Clone Rho 1D4
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Radioimmune Assays
Host Species Rabbit
Target Species Human, Mouse, Rat, Amphibian, Bovine, Zebrafish
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB015091
View Documents
Novus Biologicals
NBP2810590.025MG
0.025 mg
Each for $156.50
Add to cart
 
NB015092
View Documents
Novus Biologicals
NBP2810590.2MG
0.2 mg
Each for $419.00
Add to cart
 
Description

Description

Rhodopsin Monoclonal antibody specifically detects Rhodopsin in Human, Mouse, Rat, Amphibian, Bovine, Zebrafish Samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Radioimmunoassay.
Specifications

Specifications

Rhodopsin
Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Radioimmune Assays
Human, Mouse, Rat, Amphibian, Bovine, Zebrafish
Primary
Rho 1D4
Rabbit
Monoclonal
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit