Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RhoG Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5120669
This item is not returnable.
View return policy
Description
Immunogen sequence: TVNLNLWDTA GQEEYDRLRT LSYPQTNVFV ICFSIASPPS YENVRHKWHP EVCHHCPDVP ILLVGTKKDL RAQPDTLRRL KEQGQAPITP QQGQALAKQI H.
The Ras p21 family of guanine nucleotide proteins has been widely studied in view of its apparent role in signal transduction pathways and high frequency of mutations in human malignancies. It is now clear, however, that the Ras proteins (H-, K- and N-Ras p21) are members of a much larger superfamily of related proteins. Six members of this family, Rap 1A, Rap 1B, Rap 2, R-Ras, Ral A and Ral B, exhibit approximately 50% amino acid homology to Ras. The six mammalian Rho proteins (Rho A, B, C, G, 7 and 8) are approximately 30% homologous to Ras and are expressed in a wide range of cell types. Both Ras p21 and Rho p21, as well as other members of the Ras superfamily, contain a carboxy-terminal CAAX sequence (C, cysteine; A, aliphatic amino acid; X, any amino acid) which in the case of Ras has been shown to be essential for correct localization and function.
Specifications
RhoG | |
Polyclonal | |
Unconjugated | |
RHOG | |
2810426G09Rik; aplysia ras-related homolog G (RhoG); Arhg; ras homolog family member G; ras homolog G (RhoG); ras homolog gene family, member G; ras homolog gene family, member G (rho G); Rho G; Rhog; rho-related GTP-binding protein RhoG; Sid 10750; Sid10750 | |
Rabbit | |
Affinity chromatography | |
RUO | |
308875, 391, 56212 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
2.35 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
P84095, P84096 | |
RHOG | |
A synthetic peptide corresponding to a sequence within amino acids 50-150 of human RHOG (NP_001656.2). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction