Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WRCH1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA551270
Description
The antibody detects endogenous levels of total RHOU protein.
WRCH1 Polyclonal antibody specifically detects WRCH1 in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifications
WRCH1 | |
Polyclonal | |
Unconjugated | |
Rhou | |
2310026M05Rik; AI182090; ARHU; CDC42L1; CDC42-like GTPase 1; G28K; GTP-binding protein like 1; GTP-binding protein SB128; GTP-binding protein-like 1; hG28K; LOC678766; LOW QUALITY PROTEIN: rho-related GTP-binding protein RhoU; mG28K; ras homolog family member U; ras homolog gene family, member U; ras-like gene family member U; Ras-like protein member U; rho GTPase-like protein ARHU; Rho-related GTP-binding protein RhoU; rho-related GTP-binding protein RhoU-like; RHOU; Ryu GTPase; SB128; similar to ras homolog gene family, member U; Wnt1 responsive Cdc42 homolog; wnt-1 responsive Cdc42 homolog 1; WRCH1; WRCH-1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
58480 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
2.1 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q7L0Q8 | |
Rhou | |
Synthetic peptide of human RHOU | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction