Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RHPN1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$436.00

Specifications

Antigen RHPN1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP155003
View Documents
Novus Biologicals
NBP155003
100 μL
Each for $436.00
Add to cart
 
NBP15500320
View Documents
Novus Biologicals
NBP15500320UL
20 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

RHPN1 Polyclonal specifically detects RHPN1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

RHPN1
Polyclonal
Rabbit
Signal Transduction
Q8TCX5-2
114822
Synthetic peptides corresponding to RHPN1(rhophilin, Rho GTPase binding protein 1) The peptide sequence was selected from the middle region of RHPN1. Peptide sequence SVLFNIGALHTQIGARQDRSCTEGARRAMEAFQRAAGAFSLLRENFSHAP.
Primary
Store at -20C. Avoid freeze-thaw cycles.
Western Blot
Unconjugated
RUO
PBS and 2% Sucrose with 0.09% Sodium Azide
GTP-Rho-binding protein 1, KIAA1929outer dense fiber of sperm tails 5, ODF5, RHOPHILIN, rhophilin 1, rhophilin, Rho GTPase binding protein 1, rhophilin-1, RHPN
RHPN1
IgG
Affinity Purified
73 kDa
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit