Learn More
RHPN2 Mouse anti-Human, Clone: 1C5, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA524500
Catalog No. PIMA524500
Add to cart
Description
Description
Peptide Sequence: MTDALLPAAP QPLEKKNDGY FRKGCNPLAQ TGRSKLQNQR AALNQQILKA VRMRTGAENL LKVATNSKVR EQVRLELSFV NSDLQMLKEE LEGLNISVGV YQNTEEAFTI PLIPLGLKET KDVDFAVVLK DFILEHYSED GYLYEDEIAD LMDLRQACRT PSRDEAGVEL LMTYFIQLGF VESRFFPPTR QMGLLFTWYD SLTGVPVSQQ NLLLEKASVL FNTGALYTQI GTRCDRQTQA GLESAIDAFQ RAAGVLNYLK DTFTHTPSYD MSPAMLSVLV KMMLAQAQES VFEKISLPGI RNEFFMLVKV AQEAAKVGEV YQQLHAAMSQ APVKENIPYS WASLACVKAH HYAALAHYFT AILLIDHQVK PGTDLDHQEK CLSQLYDHMP EGLTPLATLK NDQQRRQLGK SHLRRAMAHH EESVREASLC KKLRSIEVLQ KVLCAAQERS RLTYAQHQEE DDLLNLIDAP SVVAKTEQEV DIILPQFSKL TVTDFFQKLG PLSVFSANKR WTPPRSIRFT AEEGDLGFTL RGNAPVQVHF LDPYCSASVA GAREGDYIVS IQLVDCKWLT LSEVMKLLKS FGEDEIEMKV VSLLDSTSSM HNKSATYSVG MQKTYSMICL AIDDDDKTDK TKKISKKLSF LSWGTNKNRQ KSASTLCLPS VGAARPQVKK KLPSPFSLLN SDSSWY
This gene encodes a member of the rhophilin family of Ras-homologous (Rho)-GTPase binding proteins. The encoded protein binds both GTP- and GDP-bound RhoA and GTP-bound RhoB and may be involved in the organization of the actin cytoskeleton.
Specifications
Specifications
RHPN2
Monoclonal
Unconjugated
RHPN2
Liquid
RHPN2
RHPN2 (AAH36447.1, 1 a.a. ∽ 686 a.a) full-length recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Protein A
RUO
Antibody
Monoclonal
Human
ELISA, Western Blot
1C5
PBS with no preservative; pH 7.4
Q8IUC4
RHPN2
Mouse
IgG2a, kappa
100 μg
-20° C
Primary
85415