Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RHPN2 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA5142161
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:32,000.
This gene encodes a member of the rhophilin family of Ras-homologous (Rho)-GTPase binding proteins. The encoded protein binds both GTP- and GDP-bound RhoA and GTP-bound RhoB and may be involved in the organization of the actin cytoskeleton.
Specifications
RHPN2 | |
Polyclonal | |
Unconjugated | |
RHPN2 | |
1300002E07Rik; 76 kDa RhoB effector protein; AA536890; C85860; D7Ertd784e; GTP-Rho-binding protein 2; p76RBE; RHOBP; rhophilin 2; rhophilin Rho GTPase binding protein 2; rhophilin, Rho GTPase binding protein 2; rhophilin-2; rhophilin-like Rho-GTPase binding protein; RHPN2 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
85415 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q8IUC4 | |
RHPN2 | |
Peptide with sequence C-TNKNRQKSAST. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction