Learn More
Akron Biotechnology LLCSupplier Diversity Partner RHU FGF-21 100UG DFS Item
Manufacturer:  Akron Biotechnology LLCSupplier Diversity Partner AK82160100
rHu FGF-21 is produced in E.coli is a single, non-glycosylated polypeptide chain containing 182 amino acids and having a molecular mass of 19.5 kDa. 100ug
Catalog No. 50-753-3202