Learn More
Akron Biotechnology LLCSupplier Diversity Partner RHU GALECTIN-1 10UG DFS Item
Manufacturer:  Akron Biotechnology LLCSupplier Diversity Partner AK82810010
rHu Galectin-1 controls cell growth and proliferation and induces apoptosis of activated T cells. It can also modulate cytokine secretion or inhibit pro-inflammatory cytokine production. 10ug
Catalog No. 50-753-3206