Learn More
Akron Biotechnology LLCSupplier Diversity Partner RHU GRO-BETA 2UG DFS Item
Manufacturer:  Akron Biotechnology LLCSupplier Diversity Partner AK82890002
rHu Gro-beta genes encode 3 highly related chemokines, MGSA/GRO alpha, -beta and -gamma. All 3 proteins bind to the same receptors, but with differing affinities, and stimulate a number of biological responses. 2ug
Catalog No. 50-753-3210